Anti-PLUNC Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-5913
102026-794EA 718.77 USD
102026-794
Anti-PLUNC Rabbit Polyclonal Antibody
Antibodies
PLUNC is the human homolog of murine plunc and is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this protein is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer.

WB: 0.25 ug/ml

Type: Primary
Antigen: PLUNC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR