Anti-PLK1 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-5563
102026-094EA 718.77 USD
102026-094
Anti-PLK1 Rabbit Polyclonal Antibody
Antibodies
Serine/threonine-protein kinase that performs several important functions throughout M phase of the cell cycle, including the regulation of centrosome maturation and spindle assembly, the removal of cohesins from chromosome arms, the inactivation of APC/C inhibitors, and the regulation of mitotic exit and cytokinesis.

WB: 1 ug/ml

Type: Primary
Antigen: PLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR