Anti-PLEKHH2 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-3507
102021-982EA 718.77 USD
102021-982
Anti-PLEKHH2 Rabbit Polyclonal Antibody
Antibodies
PLEKHH2 contains 1 FERM domain, 1 MyTH4 domain and 2 PH domains. It is a single-pass membrane protein. The exact function of PLEKHH2 remains unknown.

WB: 1 ug/ml

Type: Primary
Antigen: PLEKHH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR