Anti-PCDH8 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-6133
102027-234EA 718.77 USD
102027-234
Anti-PCDH8 Rabbit Polyclonal Antibody
Antibodies
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. PCDH8 is an integral membrane protein that is thought to function in cell adhesion in a CNS-specific manner. Unlike classical cadherins, which are generally encoded by 15-17 exons, this gene includes only 3 exons. Notable is the large first exon encoding the extracellular region, including 6 cadherin domains and a transmembrane region.

WB: 1 ug/ml

Type: Primary
Antigen: PCDH8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR