Anti-PCDH17 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-2580
102020-128EA 718.77 USD
102020-128
Anti-PCDH17 Rabbit Polyclonal Antibody
Antibodies
PCDH17 contains six extracellular cadherin domains, a transmembrane domain, and a cytoplasmic tail differing from those of the classical cadherins.It may play a role in the establishment and function of specific cell-cell connections in the brain.

WB: 1 ug/ml

Type: Primary
Antigen: PCDH17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR