Anti-Pcdh11x Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-8668
102032-304EA 718.77 USD
102032-304
Anti-Pcdh11x Rabbit Polyclonal Antibody
Antibodies
Pcdh11x is a potential calcium-dependent cell-adhesion protein.

Optimal conditions to be determined by the end user

Type: Primary
Antigen: Pcdh11x
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR