Anti-PCBD1 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-7922
102030-812EA 718.77 USD
102030-812
Anti-PCBD1 Rabbit Polyclonal Antibody
Antibodies
PCBD1 is involved in tetrahydrobiopterin biosynthesis. It seems to both prevent the formation of 7-pterins and accelerate the formation of quinonoid-BH2. It is a coactivator for HNF1A-dependent transcription. It regulates the dimerization of homeodomain protein HNF1A and enhances its transcriptional activity.

Optimal conditions to be determined by the end user

Type: Primary
Antigen: PCBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR