Anti-PBX2 Mouse Monoclonal Antibody [clone: 156C1_2a]

Supplier: Fitzgerald Industries
10R-1643
101979-786EA 668.31 USD
101979-786
Anti-PBX2 Mouse Monoclonal Antibody [clone: 156C1_2a]
Antibodies
This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.

ICC: 2-100 ug/ml

Type: Primary
Antigen: PBX2
Clonality: Monoclonal
Clone: 156C1_2a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR