Anti-PBP2a Mouse Monoclonal Antibody [clone: M8121521]

10-P08A
101978-392EA 338.46 USD
101978-392
Anti-PBP2a Mouse Monoclonal Antibody [clone: M8121521]
Penicillin-binding proteins (PBPs) are a group of proteins that are characterized by their affinity for and binding of penicillin. They are a normal constituent of many bacteria; the name just reflects the way by which the protein was discovered. All beta-lactam antibiotics (except for tabtoxinine-Beta-lactam, which inhibits glutamine synthetase) bind to PBP to have their effect of preventing cell wall construction by the bacterium.

Optimal conditions to be determined by end user

Type: Primary
Antigen: PBP2a
Clonality: Monoclonal
Clone: M8121521
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Staphylococcus aureus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR