Anti-PARP2 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-2437
102019-842EA 718.77 USD
102019-842
Anti-PARP2 Rabbit Polyclonal Antibody
Antibodies
PARP2 contains a catalytic domain and is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. The basic residues within the N-terminal region of this protein may bear potential DNA-binding properties, and may be involved in the nuclear and/or nucleolar targeting of protein.

WB: 1.6 ug/ml

Type: Primary
Antigen: PARP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR