Polyethylene Tubing, ALA Scientific Instruments
Supplier: ALA Scientific Instruments
PET-20
100133-852EA 95.63 USD
100133-852
Polyethylene Tubing, ALA Scientific Instruments
Tubing PE Tubing
This chemically inert, non-contaminating, translucent tubing is ideal for laboratory, hospital, food and beverage applications.
  • A low-density polyethylene produced from 100% virgin resin
  • Accepts metal and plastic compression fittings
Well suited for use with pneumatic controls, instrumentation, and hydraulic applications where its exceptional stability, long life, and resistance to corrosion and environmental chemicals enhance system realiability.
Certifications: Meets FDA standards.