Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Compact Tube Furnaces, Model RD, Nabertherm

Supplier: NABERTHERM INC.
RD30A171A-208 RD30A171A-240 RD30A173A-240 RD30A173A-208
The RD tube furnaces convince with their unbeatable price-performance ratio, very compact outer dimensions and their low weight.

  • Housing made of sheets of textured stainless steel
  • Inner diameter of the tube: 30 mm, heated length: 200 mm
  • Working tube made of C 530 material including two fiber plugs as standard
  • Thermocouple type K (1100 °C) or type S (1300 °C)
  • Solid state relays provide for low-noise operation of the heating
  • Heating wires wound directly around the working tube resulting in very fast heat-up rates

Both models are designed for horizontal application. If the customer requires protective gas atmosphere, a separate working tube incl. gas supply system 1, made of quartz glass, must be inserted in the working tube.

These all-rounders are equipped with a working tube which also serves as support for the heating wires. Thus, the working tube is part of the furnace heating which has the advantage that the furnaces achieve very high heat-up rates. The tube furnaces can be supplied for 1100 °C or 1300 °C.

Accessories information: Optional accessories include: over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; gas supply system for non-flammable protective or reactive gas. Contact your VWR representative for more information.

Ordering information: These compact tube furnaces can be ordered in a variety of maximum temperatures and voltage options. Models rated up to 1100 °C or 1300 °C. Basic control with the R7 controller that includes one set temperature.



 

ORDER

Expand All / Collapse All

Compact Tube Furnaces, 208 V
Electrical Model Heated Length Maximum Temperature Control Type Heat-Up Time Length Constant Temperature Supplier No. VWR Catalog Number Unit Price Quantity
208 V RD 30/200/11 200 mm 1100 °C R7 20 min. to Tmax 65 ± 5 K in mm3 RD30A171A-208 75994-082 Each Retrieving
208 V RD 30/200/13 200 mm 1300 °C R7 25 min. to Tmax 65 ± 5 K in mm3 RD30A173A-208 75994-086 Each Retrieving

Compact Tube Furnaces, 240 V
Electrical Model Heated Length Maximum Temperature Control Type Heat-Up Time Length Constant Temperature Supplier No. VWR Catalog Number Unit Price Quantity
240 V RD 30/200/11 200 mm 1100 °C R7 20 min. to Tmax 65 ± 5 K in mm3 RD30A171A-240 75994-084 Each Retrieving
240 V RD 30/200/13 200 mm 1300 °C R7 25 min. to Tmax 65 ± 5 K in mm3 RD30A173A-240 75994-088 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Power 1.5 KW, 1-phase
Weight 12 kg
Outside Height 350 mm
Tube int.Ø 30 mm
Outside Width 350 mm
Outside Depth 200 mm

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty