N-, Formylmethionine-secific TRNA

Supplier: MP Biomedicals
0219915410 0219915450
IC0219915450EA 3748.31 USD
IC0219915450 IC0219915410
N-, Formylmethionine-secific TRNA
Proteins and Peptides
Ribonucleic Acid, Transfer N-Formylmethionine Specific from Escherichia Coli Mre 600 Acceptor Activities Given In Picomoles (10-12 Moles) Per A260 Unit. One Unit Yields An A260 Of 1.0 In 1.0 Ml Of Water (1 Cm Light Path). Acceptor Activity: Approx. 1300 Picomoles Per A260 Unit.

Storage temperature: -20 °C

Soluble in water (50 mg/mL - clear, colorless to yellowish solution).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR