You Searched For: Scalpel Handles
High-quality scalpels are available in durable, autoclavable models for repeat procedural use or in disposable forms to provide an economical cutting tool. Scalpel handles are sold separately with some models featuring graduated measurements in millimeters and centimeters. Sterile disposable blades are ideal for cutaneous surgery such as shave biopsy, saucerization of flat lesions, and leveling of pedunculated lesions. Stainless steel models are also available for superior cutting performance and a high degree of resistance to corrosion and ease-of-use.
High-quality scalpels are available in durable, autoclavable models for repeat procedural use or in disposable forms to provide an economical cutting tool. Scalpel handles are sold separately with some models featuring graduated measurements in millimeters and centimeters. Sterile disposable blades are ideal for cutaneous surgery such as shave biopsy, saucerization of flat lesions, and leveling of pedunculated lesions. Stainless steel models are also available for superior cutting performance and a high degree of resistance to corrosion and ease-of-use.
58  results were found
SearchResultCount:"58"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR International
Description: VWR® Surgeon Grade Scalpel Blades are made of high quality surgical stainless steel.
Supplier: VWR International
Description: These high-quality scalpel knife handle fits the VWR® 10 and 20 series scalpel blades.
Supplier: VWR International
Description: These high-quality round scalpel knife handle fits the VWR® stainless and carbon steel scalpel blades.
Supplier: VWR International
Description: These high-quality scalpel knife handle fits the VWR® 10 and 20 series scalpel blades.
Supplier: VWR International
Description: These high-quality autopsy scalpel knife handle fits the VWR® 60 and 70 scalpel blades.
Catalog Number: (10170-208)
Supplier: Epredia
Description: Securely hold blades with the Shandon™ nickel-plated scalpel blade handle, constructed of rustless, nickel-plated steel #4.
Supplier: Sklar
Description: Merit™ Scalpel Handles are stainless steel, industry standard.
Supplier: Sklar
Description: Sklar's Scalpel Handles are compatible with disposable blades and used for sharp dissection
Supplier: Miltex
Description: EZ-OFF reusable plastic knife handles.
Supplier: Miltex
Description: Handles feature mm and cm graduations
Supplier: Sklar
Description: Econo™ Surgical Knife Handles are Stainless Steel and Industry Standard.
Catalog Number: (95053-138)
Supplier: Sklar
Description: Sklar's Scalpel Handle is straight, reusable, and made from stainless steel.
Catalog Number: (76192-330)
Supplier: Sklar
Description: Econo™ Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection.
Supplier: Sklar
Description: Sklar's Large Lab Type Scalpel Handles are designed for use in sharp dissection
Supplier: Miltex
Description: Fits surgical blades numbers 10-15c
Catalog Number: (95053-140)
Supplier: Sklar
Description: Sklar's #3K Scalpel Handle is a round, knurled, beaver-style handle used for holding microsurgical series 50, 60, and 70 scalpel blades
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 58
no targeter for Bottom