TestStrip™ Test Strips, Glucose, BioVision
Supplier: BioVision
TestStrip™
M1093-100 M1093-200
76005-312EA 180.96 USD
76005-312 76005-310
TestStrip™ Test Strips, Glucose, BioVision
Clinical Diagnostic Systems Clinical Reagents and Kits
TestStrip™ Glucose lab strips that measures glucose concentration in mg/dL.
  • Measures glucose concentration
  • Sensitive
  • Detects glucose from breakdown of starch
TestStrip™ Glucose lab strips are semi-quantitative strips that measures glucose concentration in mg/dL and are specific to glucose (0.1 – 0.01%).
The strips are sensitive enough to detect the glucose from the breakdown of starch.