    Sort by:
  • Search Within Results
Amphibian ringer solution
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Electron Microscopy Sciences
Description:   Amphibian Ringer solution ingredients are NaCl; KCl; CaCl₂·6H₂O and distilled water.
Minority or Woman-Owned Business Enterprise