CRAC Channel Inhibitor, BTP2, Calbiochem®
Supplier: MilliporeSigma
Synonyms:
N-(4-(3,5-Bis(trifluoromethyl)-1H-pyrazol-1-yl)phenyl)-4-methyl-1,2,3-thiadiazole-5-carboxamide
Packaged under inert gas.
203890-5MG
82603-704EA
237.31
USD
82603-704
CRAC Channel Inhibitor, BTP2, Calbiochem®
CRAC Channel Inhibitor, BTP2
A cell-permeable pyrazole that acts as a more potent blocker of Stim1 and Orai1-coupled CRAC channel-mediated SOCE (store-operated Ca2+ entry) in RBL-2H3 cells.
- Solid form
- Complement to Pyr 3,Pyr6, and Pyr10 in Ca2+ signaling studies
Formula:
C₁₅H₉F₆N₅OS MW: 421.32 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00220976 CAS Number: 223499-30-7 |
Specification Test Results
Primary Target | CRAC Channel |
Primary Target IC50 | 590 nM by Fura-2 |
Purity | ≥95% by HPLC |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...