Pacific Hemostasis® Thrombin Time Reagent, Thermo Scientific
Supplier: Thermo Scientific
100011
89217-252EA 72.26 USD
89217-252
Pacific Hemostasis® Thrombin Time Reagent, Thermo Scientific
Thrombin Time Reagent is used to screen for conditions that can interfere with the conversion of fibrinogen to fibrin
It is prepared from bovine source material with buffers.