SP Wilmad-LabGlass CHROMWARE® Economy HPLC Mobile Phase Filtration/Degassing Systems, SP Industries
BP-1975-002 BP-1975-020 BP-1975-001 BP-1975-005 BP-1975-010
14207-954EA 1178.43 USD
14207-954 14205-668 14206-106 14206-132 14207-676
SP Wilmad-LabGlass CHROMWARE® Economy HPLC Mobile Phase Filtration/Degassing Systems, SP Industries
The economy HPLC mobile phase filtration/degassing system is offered for those chromatographers who want the benefits of filtering directly into a CHROMWARE® HPLC reservoir, but don't need the air-tight three-hole cap.
Ordering information: Complete economy HPLC mobile phase filtration/degassing system consists of:
BP-1901 TORNADO® HPLC Reservoir with Red UV Safety Coating (qty 1)
BP-2310-118 TORNADO® PTFE Stirrer (qty 1)
BP-1915-047 47mm Filtration/Degassing Cap (qty 1)
BP-1904-048 47mm Mobile Phase Pickup Adapter w/Valve (qty 1)