Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potter-Elvejhem Tissue Grinders with PTFE Pestle, Wilmad-LabGlass

Supplier: WILMAD/LABGLASS
LG-10651-102 LG-10651-100 LG-10651-010
These Potter-Elvehjem tissue grinders have a PTFE pestle attached to a stainless steel shaft with precision formed, unground mortar.

  • They are used in preparing small samples for pathological and biochemical studies
  • Tissue grinder standard clearance is 0.004 to 0.006"

Homogenization occurs as the sample and buffer are forced through the cylindrical mortar and the pestle is rotated downward.

Ordering information: Other clearances can be obtained via special order.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Capacity Shaft Diameter Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mL 3/16" LG-10651-010 14212-836 Each Retrieving
1 mL 3/16" LG-10651-100 14212-826 Each Retrieving
3 mL 3/16" LG-10651-102 14212-832 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty