Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potter-Elvejhem Glass Tissue Grinder with Glass Pestle, Wilmad-LabGlass

Supplier: WILMAD/LABGLASS
LG-10650-100
This Potter-Elvehjem tissue grinder is used in preparing small samples for pathological and biochemical studies.

  • Homogenization occurs as the sample and buffer are forced through the cylindrical mortar and the pestle is rotated downward
  • Mortar and pestle are fabricated from borosilicate glass with finely ground contact surfaces

If used with a motor-driven device, use flexible tubing as a friction clutch to the stirrer motor to prevent damage to the apparatus. Tissue grinder standard clearance is 0.004" to 0.006”.

Ordering information: Other clearances can be obtained via special order.



 

ORDER

Expand All / Collapse All
Capacity Shaft Diameter Supplier No. VWR Catalog Number Unit Price Quantity
5 mL 6 mm LG-10650-100 14208-248 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty