SP Wilmad-LabGlass S Connectors, SP Industries
Supplier: SP Industries
LG-4644-100
14008-188EA 28.15 USD
14008-188
SP Wilmad-LabGlass S Connectors, SP Industries
These S Connectors feature separate adjustment capability when used with two rods or clamps, as opposed to the single clamp action of the Hook Connector.
  • Clamp items at a 90° angle