SP Wilmad-LabGlass Power Hold Universal Clamp, SP Industries
Supplier: SP Industries
LG-9571-100
14009-530EA 133.84 USD
14009-530
SP Wilmad-LabGlass Power Hold Universal Clamp, SP Industries
Clamps General Purpose Clamps
This Universal Clamp is constructed of precision machined aluminum for strength and long life.
  • Holds any stirrer, with mounting rod from 3/8" to 5/8" diameter, with precision adjustment
  • Easy to slide up and down
  • Large knob provides power to clamp securely
  • Features adjustable stop for safety and speed in operating and setup