Cytidine Cell Culture Reagent
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one
Cytidine is a pyrimidine nucleoside that is composed of the base cytosine linked to the five-carbon sugar d-ribose. There are a variety of cytidine analogs with potentially useful pharmacology .
Cytidine when intraperitoneal administered in combination with choline improves nerve regeneration and functional recovery in a rat model of sciatic nerve injury. Used as an ex vivo drug selectable marker in gene-modified primary bone marrow stromal cells for cytidine deaminase assay.
Physical Appearance: White Powder
Optical Rotation: +34.6 ° (Lit.)
UV/Visible Absorbance: λ max (water): 270 - 272 nm
Solubility: Soluble in water (50 mg/mL- clear colorless to faint yellow solution)
Formula:
C₉H₁₃N₃O₅ MW: 243.22 g/mol Melting Pt: 208…218 °C |
MDL Number:
MFCD00006545 CAS Number: 65-46-3 EINECS: 200-610-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...