Salicylaldehyde azine

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: SalazineBis(2-hydroxybenzaldehyde) hydrazoneSalicylaldazine2,2'-(Hydrazine-1,2-diylidenebis(methanylylidene))diphenolN,N'-Bis(salicylidene)hydrazine

0521809080
IC0521809080EA 1101.26 USD
IC0521809080
Salicylaldehyde azine
Salicylaldehyde azine

Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
Physical Appearance: Yellow Powder

Formula: C₁₄H₁₂N₂O₂
MW: 240.26 g/mol
Melting Pt: 213…220 °C
MDL Number: MFCD00043496
CAS Number: 959-36-4
EINECS: 213-500-0
https://us.vwr.com/stibo/web/std.lang.all/01/44/36440144.jpg

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Reference
Melting Point 215.1 - 217.7°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR