IGEPAL® CA-630
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Octylphenyl-polyethylene glycol
0219859650
0219859680
0219859690
IC0219859680EA
91.08
USD
IC0219859680
IC0219859690
IC0219859650
IGEPAL® CA-630
IGEPAL® CA-630
IGEPAL® CA-630 a homologous series of octylphenoxypoly(ethyleneoxy)ethanols. Chemically indistinguishable from Nonidet P-40, which is no longer available.
- For solubilizing and isolation of membrane proteins
Physical Appearance: Clear, Colorless, Viscous, Liquid
Viscosity: 250-350 cP, neat(25 °C)(lit.)
Color APHA: ≤100
Critical Micelle Concentration: 0.08 mM(20-25°C)
Formula:
(C₂H₄O)nC₁₄H₂₂O Boiling Pt: >200 °C (lit.) Melting Pt: +4 °C (lit.) Density: 1.06 g/cm³ (25 °C) (lit.) Flash Pt: 251 °C (483.8 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00132506 CAS Number: 9036-19-5 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...