HALO-5® RP-Amide 5 micron Fused-Core® HPLC Columns, USP-L60, Mac Mod
95814907 98515607 95810507 95813607 95810607 95813407 98518707 95811307 95812407 95813707 95811507 98519707 95810407 98519607 95812207 98515707 95812907 95811907 98517607 95812607 95812507 95811707 98516707 98517407 98518607 95813507 98517707 95813907 98516607 98515407 95814507 95811607 98519407 98518407 95814407 95814307 95813307 95813207 95812307 95814207 98516407 95810707 95811407 95812707
102969-938EA 1399.15 USD
102969-938 102969-718 102969-816 102970-050 102969-814 102969-758 102969-812 102969-954 102969-778 102969-756 102969-810 102969-754 102969-732 102969-478 102969-552 102970-002 102969-794 102969-970 102969-210 102969-792 102969-494 102969-692 102969-790 102970-066 102969-508 102969-706 102969-804 102970-082 102969-768 102969-746 102969-788 102969-986 102969-786 102969-522 102969-764 102969-762 102969-784 102969-266 102969-760 102970-114 102970-098 102970-130 102970-034 102970-018
HALO-5® RP-Amide 5 micron Fused-Core® HPLC Columns, USP-L60, Mac Mod
Fused-Core particle technology produces UHPLC columns that provide ultrahigh resolution and ultrafast separations without sacrificing column ruggedness and reliability
  • For small molecule separations
  • L60 USP designation
  • Complementary selectivity to alkyl phases
  • Enhanced stability for minimum bleed and long life
  • Maximum carbon load as 7.3%
HALO columns generate significantly less back pressure compared to other UHPLC columns, putting less stress on the system and facilitating rugged reliable performance. The HALO-5 columns provides comparable efficiency and resolution to 3 µm non-core columns for HPLC separations at 50% lower back pressure.