Exonuclease I (from E. coli strain containing an overproducing clone of E. coli Exonuclease I)
Supplier: BioVision
9217-500
75838-154EA 145.72 USD
75838-154
Exonuclease I (from E. coli strain containing an overproducing clone of E. coli Exonuclease I)
Enzymes
Exonuclease I hydrolyzes single-stranded DNA in the 3'→5' direction, releasing 5'-mononucleotides and leaving the terminal 5'-dinucleotide intact. Hydrolysis is processive and cannot proceed if the 3' terminus is phosphorylated.