Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human CellExp™ Biotinylated PD-1 / PDCD1, His Tag & Fc Tag, human recombinant, BioVision.Inc

Supplier: BioVision
7877-5
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway.
The Biotinylated Human PD-1 / PDCD1 Protein, His Tag & Fc Tag was produced using proprietary single-point enzymatic biotinlabeling technology at C-terminus to retain its native conformational structure and, therefore, bioactivity, and has been used successfully for high sensitivity and low variability assays, such as ELISA, western blotting, inhibition assay, pull-down, SPR, FACS, and affinity purification, together with anti-biotin antibody, streptavidin or avidin-conjugated probes.




 

ORDER

Expand All / Collapse All
Size Biological Activity Supplier No. VWR Catalog Number Unit Price Quantity
5 μg Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1, Fc Tag at 5 μg/mL (100 µl/well), can bind Biotinylated Human PD-1, His Tag & Fc Tag with a linear range of 0.01-0.1 μg/mL. 7877-5 75838-032 Each (5µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Conjugation Unconjugated
Gene ID Q15116
Molecular Weight Biotinylated rhPDCD1 Fc Chimera, fused with Fc fragment of human IgG1 at the C-terminus, has a calculated MW of 42.6 kDa. The predicted N-terminus is Pro 21. DTT-reduced Protein migrates as 50 kDa due to glycosylation. The primary amine of biotinylated rhPDCD1 Fc Chimera was labeled by biotin for detection in ELISA, dot blot or western blot using streptavidin or avidin-conjugated probes.
Shipping Temperature Gel Pack
Source HEK293 cells
Storage Conditions -20°C
Protein PD-1/PDCD1
Endotoxin Content <1 EU/μg by LAL method
Species Human
Reconstitution Instructions Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 µg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.
Protein Type Recombinant
Protein Synonyms CD279, hPD-1, SLEB2, PD1, PDCD1, hPD-l

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty