HIV2 Recombinant HIV-2 Protease (from E. coli), BioVision
Supplier: BioVision
7851-100 7851-20
75837-980EA 239.32 USD
75837-980 75837-982
HIV2 Recombinant HIV-2 Protease (from E. coli), BioVision
Enzymes
HIV-2 Protease, an aspartyl protease (retropepsin), is essential for the life-cycle of HIV-2 sub-type virus. It is expressed in the infected cells as a part of Gag-Pol polyprotein from which it is auto-catalytically released after formation of an immature viral particle.
  • Accession number: Q68HL0
The enzyme subsequently cleaves the other parts of viral polyproteins resulting in the maturation of the virus. In HIV-infected patients, the enzyme is subjected to intensive mutagenesis and mutants resistant to applied medicines are produced as a result of the selection pressure. The mutation of HIV protease's active site or inhibition of its activity disrupts HIV’s ability to replicate and infect additional cells. HIV-2 has been found to be less pathogenic than HIV-1. The mechanism of HIV-2 is not clearly defined, nor the difference from HIV-1, however the transmission rate is much lower in HIV-2 than HIV-1.