DiaEasy™ Dialyzer (800 µl) Floating racks, Biovision Inc.
Supplier: BioVision
1018-5 1018-10
10002-978PK 81.72 USD
10002-978 10002-980
DiaEasy™ Dialyzer (800 µl) Floating racks, Biovision Inc.
Balance dialysis tubes at 2 different heights for buffer exchange during dialysis.
  • Convenient, sturdy floating racks to balance dialysis tubes at 2 different heights for buffer exchange during dialysis
Biovision’s DiaEasy™ dialyzer (800 µl) floating racks are designed to support our DiaEasy™ Dialyzer tubes of 800 µl volume. Each of these racks can hold upto 4 tubes.