NIAD-4 ≥98% (by HPLC)

Warning

Synonyms: 2-[[5'-(4-hydroxyphenyl)[2,2'-bithiophen]-5-yl]-methylene]-propanedinitrile2-[[5-[5-(4-hydroxyphenyl)thiophen-2-yl]thiophen-2-yl]methylidene]propanedinitrile

2710-5 2710-25
76021-512EA 499.82 USD
76021-512 76021-510
NIAD-4 ≥98% (by HPLC)
NIAD-4

NIAD-4 is a novel class of fluorescent probe, having good blood-brain barrier permeability, specificity to amyloid-β plaques, and "turn-on" bright far-red emission upon Aβ binding


NIAD-4 has a structure geometry that is readily planarized and results in a red shift upon binding to Aβ. The highly polarizable bridge provides a substantial bathochromic shift in both absorption and emission spectra of the agent. 

Formula: C₁₈H₁₀N₂OS₂
MW: 334.41 g/mol
Storage Temperature: Freezer
CAS Number: 868592-56-7
https://us.vwr.com/stibo/web/std.lang.all/22/28/23492228.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR