Streptozocin

Supplier: BioVision

Synonyms: Streptozotocin

AB142155-500MG AB142155-100MG
76020-804EA 207.09 USD
76020-804 76020-802
Streptozocin
Streptozocin

An antitumor antibiotic


Acts as a DNA alkylating agent that enters cells exclusively via the GLUT2 glucose transport protein. Also acts as a diabetagen,and is especially toxic to pancreatic islet insulin-producing β-cells. It is toxic to GLUT2 positive neuroendocrine tumor cells. Streptozocin has been used to induce Type 1 diabetes for medical research.

Formula: C₈H₁₅N₃O₇
MW: 265.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00006607
CAS Number: 18883-66-4
https://us.vwr.com/stibo/web/std.lang.all/20/39/23492039.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR