Anti-IgG Goat Polyclonal Antibody

Supplier: Tonbo Biosciences
70-8070-M002
76040-338EA 152.08 USD
76040-338
Anti-IgG Goat Polyclonal Antibody
Antibodies
Goat anti-Rabbit IgG (H&L) - Affinity Pure, min x w/bovine, human, or mouse IgG

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR