5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide ≥98% (by HPLC)

Synonyms: 5-((4-((2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino)pyrimidin-2-yl)amino)-2-methylbenzenesulfonamidePazopanib5-((4-((2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino)pyrimidin-2-yl)amino)-2-methylbenzenesulphonamide

CHM161-0010
76003-130EA 103.68 USD
76003-130
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide ≥98% (by HPLC)
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide

Potent and selective multi-targeted receptor tyrosine kinase inhibitor


Orally available angiogenesis inhibitor targeting vascular endothelial growth factor receptor (VEGFR), platelet-derived growth factor receptor (PDGFR) and c-kit. Against VEGFR1, VEGFR2, VEGFR3, PDGFR, FGFR, c-Kit and c-Fms, it shows IC50 values of 10 nM, 30 nM, 47 nM, 84 nM, 74 nM, 140 nM and 146 nM, respectively.

Formula: C₂₁H₂₃N₇O₂S
MW: 437.53 g/mol
MDL Number: MFCD11616589
CAS Number: 444731-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR