AGK7 ≥99% (by HPLC)
Supplier: Enzo Life Sciences
CHM146-0010
CHM146-0005
76003-084EA
228.8
USD
76003-084
76003-082
AGK7 ≥99% (by HPLC)
AGK7
Selectively inhibits sirtuin 3 over sirtuins 1 and 2.
Can be used as an inactive control in experiments with AGK2.
MW:
434.3 g/mol |
CAS Number:
304896-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...