WYE-28 ≥97% (by HPLC)
Supplier: Enzo Life Sciences
CHM137-0005
CHM137-0001
76003-048EA
124.3
USD
76003-048
76003-050
WYE-28 ≥97% (by HPLC)
WYE-28
Cell-permeable, potent, selective, and ATP-competitive mTOR inhibitor.
Cell-permeable. A potent, selective, and ATP-competitive mTOR (mammalian target of rapamycin) inhibitor (IC50 = 0.08 nM). WYE-28 displays 75-fold selectivity over PI3Kα (IC50 = 6 nM).
MW:
586.6 g/mol |
CAS Number:
1062172-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...