MMP-​9 inhibitor I ≥97% (by HPLC)

Supplier: Enzo Life Sciences

CHM133-0001 CHM133-0500
76003-036EA 97.47 USD
76003-036 76003-034
MMP-​9 inhibitor I ≥97% (by HPLC)
MMP-​9 inhibitor I

Cell-permeable, potent, selective, and reversible MMP-9 inhibitor.

MDL Number: MFCD22581508
CAS Number: 1177749-58-4
https://us.vwr.com/stibo/web/std.lang.all/16/58/45401658.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR