Syk inhibitor ≥97% (by HPLC)
Supplier: Enzo Life Sciences
CHM128-0005
76003-018EA
150.09
USD
76003-018
Syk inhibitor ≥97% (by HPLC)
Syk inhibitor
Spleen tyrosine kinase inhibitor.
Formula:
C₁₈H₁₅N₃O₃S |
MDL Number:
MFCD07776820 CAS Number: 622387-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...