SANT-1 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: N-(4-Benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine

CHM120-0005 CHM120-0001
76002-992EA 138.89 USD
76002-992 76002-990
SANT-1 ≥99% (by HPLC)
SANT-1

Potent, cell-permeable antagonist of the Shh signaling pathway.


Equipotently inhibits the activities of both wild-type and oncogenic Smoothened.

Formula: C₂₃H₂₇N₅
MW: 373.5 g/mol
MDL Number: MFCD01827306
CAS Number: 304909-07-7
https://us.vwr.com/stibo/web/std.lang.all/16/45/45401645.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR