SecinH3 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: N-(4-(5-(Benzo[d][1,3]dioxol-5-yl)-3-methoxy-1H-1,2,4-triazol-1-yl)phenyl)-2-(phenylthio)acetamide

CHM114-0005 CHM114-0010
76002-494EA 158.43 USD
76002-494 76002-496
SecinH3 ≥99% (by HPLC)
SecinH3

Cell-permeable cytohesin family small GEF antagonist.


A Sec7 domain-binding selective antagonist against cytohesin family small GEFs. SecinH3 inhibits large GEFs only at much higher concentrations and is shown to effectively inhibit insulin signaling.

Formula: C₂₄H₂₀N₄O₄S
MW: 460.5 g/mol
MDL Number: MFCD10687099
CAS Number: 853625-60-2
https://us.vwr.com/stibo/web/std.lang.all/16/39/45401639.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR