PDGF receptor tyrosine kinase inhibitor IV ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: RWJ 540973N-(3-fluorophenyl)-2,4-dihydro-6,7-dimethoxy-Indeno[1,2-c]pyrazol-3-amineJNJ-101984093-Fluoro-N-(6,7-dimethoxy-2,4-dihydroindeno[1,2-c]pyrazol-3-yl)phenylamine

CHM111-0010 CHM111-0005
76002-482EA 119.12 USD
76002-482 76002-484
PDGF receptor tyrosine kinase inhibitor IV ≥99% (by HPLC)
PDGF receptor tyrosine kinase inhibitor IV

Cell-permeable PDGFR inhibitor with antiproliferative properties.


A cell-permeable compound acting as an ATP-competitive and reversible inhibitor of PDGFR. PDGF receptor tyrosine kinase inhibitor IV displays potent antiproliferative properties in several human tumor cell lines.

Formula: C₁₈H₁₆FN₃O₂
MW: 325.34 g/mol
CAS Number: 627518-40-5
https://us.vwr.com/stibo/web/std.lang.all/16/36/45401636.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR