Erk inhibitor ≥99% (by HPLC)

Supplier: Enzo Life Sciences

CHM109-0005 CHM109-0010
76002-970EA 146.98 USD
76002-970 76002-972
Erk inhibitor ≥99% (by HPLC)
Erk inhibitor

Cell-permeable and reversible Erk inhibitor.


Displays anti-proliferative properties.

Formula: C₁₄H₁₆N₂O₃S·HCl
MW: 328.8 g/mol
CAS Number: 1049738-54-6
https://us.vwr.com/stibo/web/std.lang.all/16/34/45401634.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR