QS11 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-WN117-0025 BML-WN117-0005
76002-256EA 181.09 USD
76002-256 76002-258
QS11 ≥98% (by HPLC)
QS11

Wnt pathway activator


Synergizers with Wnt3a to activate the Wnt/β-catenin pathway. Binds and inhibits GTPase activating protein of ADP-ribosylation factor 1 (ARFGAP1), thus activating ARF. It has also been suggested that a possible mechanism by which QS11 synergizes Wnt is through ARFGAP1 inhibition leading to increased β-catenin translocation via ARF activation.

Formula: C₃₅H₃₃N₅O₂
MW: 567.7 g/mol
MDL Number: MFCD18086864
CAS Number: 944328-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR