CCT036477 ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-WN114-0005
BML-WN114-0025
76002-248EA
125.45
USD
76002-248
76002-250
CCT036477 ≥98% (by TLC)
CCT036477
Disrupts Wnt signaling
Inhibits Wnt signaling via blocking transcription at the β-catenin level independent of CBP or p300. Inhibited growth in several cancer cell lines (HT29, HCT116, SW480, and SNU475). CCT036447 was also active in vivo where it blocked development of zebrafish and Xenopus embryos and the expression of Wnt target genes.
MW:
347.8 g/mol |
CAS Number:
305372-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...