IM-12 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 3-((4-Fluorophenethyl)amino)-1-methyl-4-(2-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione

BML-WN102-0025 BML-WN102-0005
76002-410EA 117 USD
76002-410 76002-412
IM-12 ≥98% (by HPLC)
IM-12

GSK-3β inhibitor (IC50=53nM (enzymatic), 3.8µM (cellular)). The compound significantly increases β-catenin levels, leads to TCF-induction and promotes neuronal differentiation in human neural progenitor cells via activation of canonical Wnt signaling.

Formula: C₂₂H₂₀FN₃O₂
MW: 377.4115 g/mol
MDL Number: MFCD20527313
CAS Number: 1129669-05-1
https://us.vwr.com/stibo/web/std.lang.all/15/67/45401567.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR