Tubacin ≥96%

Supplier: Enzo Life Sciences

Synonyms: rel-N1-(4-((2R,4R,6S)-4-(((4,5-Diphenyloxazol-2-yl)thio)methyl)-6-(4-(hydroxymethyl)phenyl)-1,3-dioxan-2-yl)phenyl)-N8-hydroxyoctanediamide

BML-GR362-0100 BML-GR362-0500
76002-372EA 575.81 USD
76002-372 76002-370
Tubacin ≥96%
Tubacin

HDAC inhibitor


Synthetic.

Tubacin (tubulin acetylation inducer) is a highly potent and selective, reversible, cell-permeable inhibitor of HDAC6 (IC50=0.004µM). IC50's for the other HDACs are 1000-fold higher, making tubacin both more selective and more potent than Tubastatin A, which also inhibits HDAC8. Concentration in cell culture experiments typically ranges from 2-50µM.


Synthetic.


Tubacin (tubulin acetylation inducer) is a highly potent and selective, reversible, cell-permeable inhibitor of HDAC6 (IC50=0.004µM). IC50's for the other HDACs are 1000-fold higher, making tubacin both more selective and more potent than Tubastatin A, which also inhibits HDAC8. Concentration in cell culture experiments typically ranges from 2-50µM.

Formula: C₄₁H₄₃N₃O₇S
MW: 721,86 g/mol
MDL Number: MFCD28144524
CAS Number: 537049-40-4
https://us.vwr.com/stibo/web/std.lang.all/12/75/45401275.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR