Eicosapentaenoyl ethanolamide ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-FA047-0025
76002-110EA 246.54 USD
76002-110
Eicosapentaenoyl ethanolamide ≥98% (by TLC)
Eicosapentaenoyl ethanolamide
Synthetic.
Synthetic.
Has been shown to inhibit dietary-restriction-induced lifespan extension in wild-type and TOR pathway mutant nematodes. This data indicates that reduced N-acylethanolamide signaling mediates some of the effect of dietary-restriction-induced lifespan extension and that EPEA, specifically, acts as a metabolic signal that couples nutrient availability with growth and lifespan. EPEA has also been shown to have anti-proliferative effects and anti-inflammatory properties. Compound is subject to oxidation. Protect from air.
Has been shown to inhibit dietary-restriction-induced lifespan extension in wild-type and TOR pathway mutant nematodes. This data indicates that reduced N-acylethanolamide signaling mediates some of the effect of dietary-restriction-induced lifespan extension and that EPEA, specifically, acts as a metabolic signal that couples nutrient availability with growth and lifespan. EPEA has also been shown to have anti-proliferative effects and anti-inflammatory properties. Compound is subject to oxidation. Protect from air.
CAS Number: 109001-03-8
https://us.vwr.com/stibo/web/std.lang.all/38/57/45403857.jpg