SU11652 ≥98% (by HPLC)
Supplier: Enzo Life Sciences
BML-EI408-0001
76002-104EA
261.88
USD
76002-104
SU11652 ≥98% (by HPLC)
SU11652
VEGFR inhibitor
Potent, ATP-competitive and cell permeable inhibitor of tyrosine kinases (VEGF-R2 IC50=27nM, PDGF-Rβ IC50=3nM). Also inhibits wild-type and all mutant forms of Kit in mast cells (IC50's=10 to 100nM) resulting in cell cycle arrest and apoptosis. Su11652 was also shown to induce autophagy in MCF-7 cells.
MW:
414.9 g/mol |
CAS Number:
326914-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...