Tesmilifene ≥99% (by TLC)

BML-EI401-0010 BML-EI401-0050
76002-086EA 438.68 USD
76002-086 76002-084
Tesmilifene ≥99% (by TLC)
Tesmilifene

Antagonist of intracellular histamine


Tesmilifene is a novel agent that augments cytotoxicity of various chemotherapeutic agents both in vitro and in vivo. It binds selectively to the high-affinity microsomal antiestrogen binding site (Ki=50nm) but has no affinity for estrogen receptors. Inhibits concanavalin-A-induced histamine release in mast cells and acts as a novel antagonist of intracellular histamine.

Formula: C₁₉H₂₅NO·HCl
MW: 283.4 · 36.5 g/mol
CAS Number: 92981-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR